2-{2-[4-(2-Methoxyethyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049746-500MG 049746-2.5G
101888-884EA 364.62 USD
101888-884 101888-886
2-{2-[4-(2-Methoxyethyl)phenoxy]ethyl}piperidine hydrochloride
2-{2-[4-(2-Methoxyethyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO₂
MDL Number: MFCD13560148
CAS Number: 1220034-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR