3-{2-[4-(2-Methoxyethyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049745-500MG 049745-2.5G
101888-880EA 364.62 USD
101888-880 101888-882
3-{2-[4-(2-Methoxyethyl)phenoxy]ethyl}piperidine hydrochloride
3-{2-[4-(2-Methoxyethyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO₂
MDL Number: MFCD13560147
CAS Number: 1219956-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR