Ethyl-2-(3-pyrrolidinylmethoxy)benzoate hydrochloride

Supplier: Matrix Scientific

049743-500MG 049743-2.5G
101888-872EA 364.62 USD
101888-872 101888-874
Ethyl-2-(3-pyrrolidinylmethoxy)benzoate hydrochloride
Ethyl-2-(3-pyrrolidinylmethoxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO₃
MDL Number: MFCD13560145
CAS Number: 1220029-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR