Ethyl-2-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride

Supplier: Matrix Scientific

049742-500MG 049742-2.5G
101888-868EA 364.62 USD
101888-868 101888-870
Ethyl-2-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride
Ethyl-2-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MDL Number: MFCD13560144
CAS Number: 1220016-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR