Ethyl-2-[2-(3-piperidinyl)ethoxy]benzoate hydrochloride

Supplier: Matrix Scientific

049741-500MG 049741-2.5G
101888-864EA 364.62 USD
101888-864 101888-866
Ethyl-2-[2-(3-piperidinyl)ethoxy]benzoate hydrochloride
Ethyl-2-[2-(3-piperidinyl)ethoxy]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MW: 313.83 g/mol
MDL Number: MFCD13560143
CAS Number: 1220034-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR