5-Chloro[1,1'-biphenyl]-2-yl-3-pyrrolidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

049739-500MG 049739-2.5G
101888-856EA 364.62 USD
101888-856 101888-858
5-Chloro[1,1'-biphenyl]-2-yl-3-pyrrolidinylmethyl ether hydrochloride
5-Chloro[1,1'-biphenyl]-2-yl-3-pyrrolidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₉Cl₂NO
MW: 324.25 g/mol
MDL Number: MFCD13560141
CAS Number: 1220030-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR