2-{2-[(5-Chloro[1,1'-biphenyl]-2-yl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049738-500MG 049738-2.5G
101888-852EA 364.62 USD
101888-852 101888-854
2-{2-[(5-Chloro[1,1'-biphenyl]-2-yl)oxy]ethyl}piperidine hydrochloride
2-{2-[(5-Chloro[1,1'-biphenyl]-2-yl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₃Cl₂NO
MDL Number: MFCD13560140
CAS Number: 1219956-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR