3-{[2-Chloro-4-(1-methyl-1-phenylethyl)phenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049735-500MG 049735-2.5G
101888-840EA 364.62 USD
101888-840 101888-842
3-{[2-Chloro-4-(1-methyl-1-phenylethyl)phenoxy]methyl}pyrrolidine hydrochloride
3-{[2-Chloro-4-(1-methyl-1-phenylethyl)phenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₅Cl₂NO
MDL Number: MFCD13560137
CAS Number: 1220029-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR