2-Chloro-4-(1-methyl-1-phenylethyl)phenyl-2-(2-piperidinyl)ethyl ether hydrochloride
Supplier: Matrix Scientific
049734-500MG
049734-2.5G
101888-836EA
364.62
USD
101888-836
101888-838
2-Chloro-4-(1-methyl-1-phenylethyl)phenyl-2-(2-piperidinyl)ethyl ether hydrochloride
2-Chloro-4-(1-methyl-1-phenylethyl)phenyl-2-(2-piperidinyl)ethyl ether hydrochloride
Hazard: Irritant
Formula:
C₂₂H₂₉Cl₂NO MW: 394.39 g/mol |
MDL Number:
MFCD13560136 CAS Number: 1219979-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...