2-Chloro-4-(1-methyl-1-phenylethyl)phenyl-2-(3-piperidinyl)ethyl ether hydrochloride
Supplier: Matrix Scientific
049733-500MG
049733-2.5G
101888-832EA
364.62
USD
101888-832
101888-834
2-Chloro-4-(1-methyl-1-phenylethyl)phenyl-2-(3-piperidinyl)ethyl ether hydrochloride
2-Chloro-4-(1-methyl-1-phenylethyl)phenyl-2-(3-piperidinyl)ethyl ether hydrochloride
Hazard: Irritant
Formula:
C₂₂H₂₉Cl₂NO |
MDL Number:
MFCD13560135 CAS Number: 1219956-95-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...