3-{[2-(sec-Butyl)-4-chlorophenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049731-500MG 049731-2.5G
101888-824EA 364.62 USD
101888-824 101888-826
3-{[2-(sec-Butyl)-4-chlorophenoxy]methyl}pyrrolidine hydrochloride
3-{[2-(sec-Butyl)-4-chlorophenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560133
CAS Number: 1220030-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR