3-{2-[2-(sec-Butyl)-4-chlorophenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049729-500MG 049729-2.5G
101888-816EA 364.62 USD
101888-816 101888-818
3-{2-[2-(sec-Butyl)-4-chlorophenoxy]ethyl}piperidine hydrochloride
3-{2-[2-(sec-Butyl)-4-chlorophenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₇Cl₂NO
MDL Number: MFCD13560131
CAS Number: 1219979-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR