3-{[4-(tert-Butyl)-2-chlorophenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049727-500MG 049727-2.5G
101888-808EA 364.62 USD
101888-808 101888-810
3-{[4-(tert-Butyl)-2-chlorophenoxy]methyl}pyrrolidine hydrochloride
3-{[4-(tert-Butyl)-2-chlorophenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560129
CAS Number: 1219979-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR