4-{2-[4-(tert-Butyl)-2-chlorophenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049724-500MG 049724-2.5G
101888-796EA 364.62 USD
101888-796 101888-798
4-{2-[4-(tert-Butyl)-2-chlorophenoxy]ethyl}piperidine hydrochloride
4-{2-[4-(tert-Butyl)-2-chlorophenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₇Cl₂NO
MDL Number: MFCD13560126
CAS Number: 1220033-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR