3-{[2-(tert-Butyl)-4-chlorophenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049723-500MG 049723-2.5G
101888-792EA 364.62 USD
101888-792 101888-794
3-{[2-(tert-Butyl)-4-chlorophenoxy]methyl}pyrrolidine hydrochloride
3-{[2-(tert-Butyl)-4-chlorophenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560125
CAS Number: 1146960-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR