3-{2-[2-(tert-Butyl)-4-chlorophenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049721-500MG 049721-2.5G
101888-784EA 364.62 USD
101888-784 101888-786
3-{2-[2-(tert-Butyl)-4-chlorophenoxy]ethyl}piperidine hydrochloride
3-{2-[2-(tert-Butyl)-4-chlorophenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₇Cl₂NO
MDL Number: MFCD13560123
CAS Number: 1220029-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR