4-{2-[2-(tert-Butyl)-4-chlorophenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049720-500MG 049720-2.5G
101888-780EA 364.62 USD
101888-780 101888-782
4-{2-[2-(tert-Butyl)-4-chlorophenoxy]ethyl}piperidine hydrochloride
4-{2-[2-(tert-Butyl)-4-chlorophenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₇Cl₂NO
MDL Number: MFCD13560122
CAS Number: 1220030-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR