3-{[2-Chloro-4-(tert-pentyl)phenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049719-500MG 049719-2.5G
101888-776EA 364.62 USD
101888-776 101888-778
3-{[2-Chloro-4-(tert-pentyl)phenoxy]methyl}pyrrolidine hydrochloride
3-{[2-Chloro-4-(tert-pentyl)phenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅Cl₂NO
MDL Number: MFCD13560121
CAS Number: 1219979-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR