3-{2-[2-Chloro-4-(tert-pentyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049717-500MG 049717-2.5G
101888-768EA 364.62 USD
101888-768 101888-770
3-{2-[2-Chloro-4-(tert-pentyl)phenoxy]ethyl}piperidine hydrochloride
3-{2-[2-Chloro-4-(tert-pentyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₉Cl₂NO
MDL Number: MFCD13560119
CAS Number: 1220030-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR