3-{[2-Bromo-4-(tert-pentyl)phenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049715-500MG 049715-2.5G
101888-760EA 364.62 USD
101888-760 101888-762
3-{[2-Bromo-4-(tert-pentyl)phenoxy]methyl}pyrrolidine hydrochloride
3-{[2-Bromo-4-(tert-pentyl)phenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅BrClNO
MDL Number: MFCD13560117
CAS Number: 1220030-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR