3-{2-[2-Bromo-4-(tert-pentyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049713-500MG 049713-2.5G
101888-752EA 364.62 USD
101888-752 101888-754
3-{2-[2-Bromo-4-(tert-pentyl)phenoxy]ethyl}piperidine hydrochloride
3-{2-[2-Bromo-4-(tert-pentyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₉BrClNO
MDL Number: MFCD13560115
CAS Number: 1220033-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR