4-{2-[2-Bromo-4-(tert-pentyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049712-500MG 049712-2.5G
101888-748EA 364.62 USD
101888-748 101888-750
4-{2-[2-Bromo-4-(tert-pentyl)phenoxy]ethyl}piperidine hydrochloride
4-{2-[2-Bromo-4-(tert-pentyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₉BrClNO
MDL Number: MFCD13560114
CAS Number: 1219979-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR