3-[(4-Chloro-2-ethylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049711-500MG 049711-2.5G
101888-744EA 364.62 USD
101888-744 101888-746
3-[(4-Chloro-2-ethylphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Chloro-2-ethylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560113
CAS Number: 1220029-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR