2-[2-(4-Chloro-2-ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049710-500MG 049710-2.5G
101888-740EA 364.62 USD
101888-740 101888-742
2-[2-(4-Chloro-2-ethylphenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Chloro-2-ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560112
CAS Number: 1220030-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR