3-[2-(4-Chloro-2-ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049709-500MG 049709-2.5G
101888-736EA 364.62 USD
101888-736 101888-738
3-[2-(4-Chloro-2-ethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Chloro-2-ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560111
CAS Number: 1219979-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR