2-[2-(2-Chloro-4-methoxyphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049706-500MG
049706-2.5G
101888-724EA
364.62
USD
101888-724
101888-726
2-[2-(2-Chloro-4-methoxyphenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Chloro-4-methoxyphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁Cl₂NO₂ |
MDL Number:
MFCD13560108 CAS Number: 1220039-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...