3-[2-(2-Chloro-4-methoxyphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049705-500MG 049705-2.5G
101888-720EA 364.62 USD
101888-720 101888-722
3-[2-(2-Chloro-4-methoxyphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Chloro-4-methoxyphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO₂
MDL Number: MFCD13560107
CAS Number: 1220029-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR