3-[(4-Chloro-2-isopropylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049703-500MG 049703-2.5G
101888-712EA 364.62 USD
101888-712 101888-714
3-[(4-Chloro-2-isopropylphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Chloro-2-isopropylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560105
CAS Number: 1219979-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR