3-[2-(4-Chloro-2-isopropylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049701-500MG 049701-2.5G
101888-704EA 364.62 USD
101888-704 101888-706
3-[2-(4-Chloro-2-isopropylphenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Chloro-2-isopropylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅Cl₂NO
MDL Number: MFCD13560103
CAS Number: 1220030-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR