3-[(2-Chloro-4-ethylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049699-500MG 049699-2.5G
101888-696EA 364.62 USD
101888-696 101888-698
3-[(2-Chloro-4-ethylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Chloro-4-ethylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560101
CAS Number: 1220039-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR