3-[2-(2-Chloro-4-ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049697-500MG 049697-2.5G
101888-688EA 364.62 USD
101888-688 101888-690
3-[2-(2-Chloro-4-ethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Chloro-4-ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560099
CAS Number: 1219964-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR