4-[2-(2-Chloro-4-ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049696-500MG 049696-2.5G
101888-684EA 364.62 USD
101888-684 101888-686
4-[2-(2-Chloro-4-ethylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Chloro-4-ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560098
CAS Number: 1220029-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR