2-Chloro-4-methylphenyl-3-pyrrolidinylmethylether hydrochloride
Supplier: Matrix Scientific
049695-500MG
049695-2.5G
101888-680EA
364.62
USD
101888-680
101888-682
2-Chloro-4-methylphenyl-3-pyrrolidinylmethylether hydrochloride
2-Chloro-4-methylphenyl-3-pyrrolidinylmethylether hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇Cl₂NO |
MDL Number:
MFCD13560097 CAS Number: 1219956-91-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...