2-[2-(2-Chloro-4-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049694-500MG 049694-2.5G
101888-676EA 364.62 USD
101888-676 101888-678
2-[2-(2-Chloro-4-methylphenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Chloro-4-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560096
CAS Number: 1220016-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR