3-[2-(2-Chloro-4-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049693-500MG 049693-2.5G
101888-672EA 364.62 USD
101888-672 101888-674
3-[2-(2-Chloro-4-methylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Chloro-4-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560095
CAS Number: 1220030-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR