2-{2-[4-(sec-Butyl)-2-chlorophenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049690-500MG 049690-2.5G
101888-660EA 364.62 USD
101888-660 101888-662
2-{2-[4-(sec-Butyl)-2-chlorophenoxy]ethyl}piperidine hydrochloride
2-{2-[4-(sec-Butyl)-2-chlorophenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₇Cl₂NO
MDL Number: MFCD13560092
CAS Number: 1219979-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR