3-[(2-Chloro-4-isopropylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049687-500MG 049687-2.5G
101888-648EA 364.62 USD
101888-648 101888-650
3-[(2-Chloro-4-isopropylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Chloro-4-isopropylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560089
CAS Number: 1220039-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR