3-[2-(2-Chloro-4-isopropylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049685-500MG 049685-2.5G
101888-640EA 364.62 USD
101888-640 101888-642
3-[2-(2-Chloro-4-isopropylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Chloro-4-isopropylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅Cl₂NO
MDL Number: MFCD13560087
CAS Number: 1220029-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR