3-Chloro[1,1'-biphenyl]-4-yl-3-pyrrolidinylmethylether hydrochloride

Supplier: Matrix Scientific

049683-500MG 049683-2.5G
101888-632EA 364.62 USD
101888-632 101888-634
3-Chloro[1,1'-biphenyl]-4-yl-3-pyrrolidinylmethylether hydrochloride
3-Chloro[1,1'-biphenyl]-4-yl-3-pyrrolidinylmethylether hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₉Cl₂NO
MDL Number: MFCD13560085
CAS Number: 1219967-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR