2-{2-[(3-Chloro[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049682-500MG 049682-2.5G
101888-628EA 364.62 USD
101888-628 101888-630
2-{2-[(3-Chloro[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride
2-{2-[(3-Chloro[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₃Cl₂NO
MDL Number: MFCD13560084
CAS Number: 1219967-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR