4-{2-[(3-Chloro[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049680-500MG 049680-2.5G
101888-620EA 364.62 USD
101888-620 101888-622
4-{2-[(3-Chloro[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride
4-{2-[(3-Chloro[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₃Cl₂NO
MDL Number: MFCD13560082
CAS Number: 1220030-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR