3-[(2-Chloro-4,6-dimethylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049679-500MG 049679-2.5G
101888-616EA 364.62 USD
101888-616 101888-618
3-[(2-Chloro-4,6-dimethylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Chloro-4,6-dimethylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560081
CAS Number: 1220029-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR