2-[2-(2-Chloro-4,6-dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049678-500MG 049678-2.5G
101888-612EA 364.62 USD
101888-612 101888-614
2-[2-(2-Chloro-4,6-dimethylphenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Chloro-4,6-dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560080
CAS Number: 1219949-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR