3-[2-(2-Chloro-4,6-dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049677-500MG 049677-2.5G
101888-608EA 364.62 USD
101888-608 101888-610
3-[2-(2-Chloro-4,6-dimethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Chloro-4,6-dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560079
CAS Number: 1220030-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR