4-[2-(2-Chloro-4,6-dimethylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049676-500MG
049676-2.5G
101888-604EA
364.62
USD
101888-604
101888-606
4-[2-(2-Chloro-4,6-dimethylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Chloro-4,6-dimethylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₃Cl₂NO |
MDL Number:
MFCD13560078 CAS Number: 1219967-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...