3-{[4-(tert-Butyl)-2-methylphenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049675-500MG 049675-2.5G
101888-600EA 364.62 USD
101888-600 101888-602
3-{[4-(tert-Butyl)-2-methylphenoxy]methyl}pyrrolidine hydrochloride
3-{[4-(tert-Butyl)-2-methylphenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560077
CAS Number: 1220039-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR