2-{2-[(5-Bromo[1,1'-biphenyl]-2-yl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049670-500MG 049670-2.5G
101888-580EA 364.62 USD
101888-580 101888-582
2-{2-[(5-Bromo[1,1'-biphenyl]-2-yl)oxy]ethyl}piperidine hydrochloride
2-{2-[(5-Bromo[1,1'-biphenyl]-2-yl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₃BrClNO
MDL Number: MFCD13560072
CAS Number: 1219963-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR