3-{2-[(5-Bromo[1,1'-biphenyl]-2-yl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049669-500MG 049669-2.5G
101888-576EA 364.62 USD
101888-576 101888-578
3-{2-[(5-Bromo[1,1'-biphenyl]-2-yl)oxy]ethyl}piperidine hydrochloride
3-{2-[(5-Bromo[1,1'-biphenyl]-2-yl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₃BrClNO
MDL Number: MFCD13560071
CAS Number: 1219967-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR