3-{[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049667-500MG 049667-2.5G
101888-568EA 364.62 USD
101888-568 101888-570
3-{[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]methyl}pyrrolidine hydrochloride
3-{[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₅BrClNO
MDL Number: MFCD13560069
CAS Number: 1220029-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR