2-{2-[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]ethyl}piperidine hydrochloride
Supplier: Matrix Scientific
049666-500MG
049666-2.5G
101888-564EA
364.62
USD
101888-564
101888-566
2-{2-[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]ethyl}piperidine hydrochloride
2-{2-[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]ethyl}piperidine hydrochloride
Hazard: Irritant
Formula:
C₂₂H₂₉BrClNO |
MDL Number:
MFCD13560068 CAS Number: 1219967-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...